Please login to your online account to display your discounted pricing

Fisher BioReagents™ EZ-Run™ Prestained Rec Protein Ladder


Comprised of 10 recombinant proteins covalently coupled to a blue chromophore plus 10 and 72kDa reference bands tagged with green and orange dyes, respectively

Manufacturer: fisher bioreagents™  BP36031

 View more versions of this product

 Disclaimers

This product is manufactured under the license for Strep-tag™ technology covered by U.S. Patent No. 5,506,121; U.K. Patent No. 2272698; and French Patent No. 93 13 066.

Catalog No. BP36031

  • / Pack of 2 Each
 

Description & Specifications

Specifications

Storage Requirements −20°C
Range 10 to 72 kd
Formulation Supplied in gel-loading buffer
Quantity 2 × 500μL
Color Blue
Sufficient For 200 Loadings
For Use With (Application) Monitoring protein separation during SDS-PAGE