Please login to your online account to display your discounted pricing

Fisher BioReagents™ CPG Columns for Automated DNA Synthesizers


Fit most DNA synthesizers

Manufacturer: fisher bioreagents™  BP276710

 View more versions of this product

Catalog No. BP276710

  • / Pack of 10 Each
 

Description & Specifications

Specifications

For Use With (Application) DNA synthesis
Product Type Column
For Use With (Equipment) Automated DNA synthesizers
Concentration 0.2uM
Quantity 10 Pack
Concentration 0.2uM
Quantity 10 Pack