Fisher BioReagents™ CPG Columns for Automated DNA Synthesizers
Fit most DNA synthesizers
$113.57 - $113.57
Products ${productFamilyLength}
Catalog Number Mfr. No. Price Quantity  
Catalog Number Mfr. No. Price Quantity  
BP276710 Fisher BioReagents™
BP276710
Pack of 10 Each for $113.57
Specifications
Description
  • 500Å Controlled Pore Glass Columns made to fit most DNA synthesizers
  • Choice of two sizes for low- or high-volume synthesis
Note: Can use with Beckman synthesizers if adapter is used.