Please login to your online account to display your discounted pricing

Fisher BioReagents™ ChromoMax™ IPTG/X-Gal Solution


Accelerate your blue/white colony screening

Manufacturer: fisher bioreagents™  BP420050

 View more versions of this product

Catalog No. BP4200-50

  • / Each
 

Description & Specifications

Specifications

Format Liquid
Quantity 5 x 10mL
Storage Requirements -20°C
Packaging Bottle
Additive IPTG/X-Gal Solution
For Use With (Application) All commercially available competent cells that utilize lacZ blue/white screening