Please login to your online account to display your discounted pricing

Fisher BioReagents™ Base4™ DNA Synthesis Reagents


For use with DNA synthesizers

Manufacturer: fisher bioreagents™  BP3145-200

 View more versions of this product

Catalog No. BP3145200

  • / Each
  • / Case of 6 Each
 

Description & Specifications

Specifications

For Use With (Application) DNA synthesis
Quantity 200mL
Product Type Reagent
Formulation Iodine in THF/Pyridine/Water 0.02M (70:20:10)
Concentration .02M
pH <2