Please login to your online account to display your discounted pricing

Fisher BioReagents™ Base4™ DNA Synthesis Reagents


For use with DNA synthesizers

Manufacturer: fisher bioreagents™  BP3130N2-19

 View more versions of this product

Catalog No. BP3130N219

  • / Each
 

Description & Specifications

Specifications

pH <2
Product Type Reagent
Formulation N-Methylimidazole 10% in THF
Quantity 19000mL
For Use With (Application) DNA synthesis