Please login to your online account to display your discounted pricing

Fisher BioReagents™ Base4™ DNA Synthesis Reagents


For use with DNA synthesizers

Manufacturer: fisher bioreagents™  BP3120N2-19

 View more versions of this product

Catalog No. BP3120N219

  • / Each
 

Description & Specifications

Specifications

For Use With (Application) DNA synthesis
Formulation THF/Pyridine/Acetic Anhydride (8:1:1)
Product Type Reagent
Quantity 19000mL
pH <2