Please login to your online account to display your discounted pricing

Fisher BioReagents™ Base4™ DNA Synthesis Reagents


For use with DNA synthesizers

Manufacturer: fisher bioreagents™  BP3150N2-19

 View more versions of this product

Catalog No. BP3150N219

  • / Each
 

Description & Specifications

Specifications

Formulation TCA 3% in Methylene Chloride
Product Type Reagent
pH <2
For Use With (Application) DNA synthesis
Quantity 19000mL