Please login to your online account to display your discounted pricing

Fisher BioReagents™ Base4™ DNA Synthesis Reagents


For use with DNA synthesizers

Manufacturer: fisher bioreagents™  BP3150-450

 View more versions of this product

Catalog No. BP3150450

  • / Each
  • / Case of 6 Each
 

Description & Specifications

Specifications

Quantity 450mL
pH <2
For Use With (Application) DNA synthesis
Product Type Reagent
Formulation TCA 3% in Methylene Chloride