Please login to your online account to display your discounted pricing

Fisher BioReagents™ Base4™ DNA Synthesis Reagents


For use with DNA synthesizers

Manufacturer: fisher bioreagents™  BP3150-4

 View more versions of this product

Catalog No. BP31504

  • / Each
  • / Case of 4 Each
 

Description & Specifications

Specifications

pH <2
Formulation TCA 3% in Methylene Chloride
Product Type Reagent
Quantity 4000mL
For Use With (Application) DNA synthesis