missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisher BioReagents™ Base4™ DNA Synthesis Reagents

For use with DNA synthesizers

$54.11 - $2,047.78



Products ${productFamilyLength}
Catalog Number Mfr. No. Price Quantity  
Catalog Number Mfr. No. Price Quantity  
BP3120N219 Fisher BioReagents™
BP3120N2-19
Each for $1,647.51
BP3130N219 Fisher BioReagents™
BP3130N2-19
Each for $2,047.78
BP3145200 Fisher BioReagents™
BP3145-200
Each for $54.11
Case of 6 Each for $324.66
BP3150N219 Fisher BioReagents™
BP3150N2-19
Each for $1,266.84
Specifications

Description

Available in 4L amber glass bottles for high-throughput DNA synthesis.