missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Finntip™ Filtered Pipette Tips
GREENER_CHOICE

Catalog No. 21377604
Change view
Click to view available options
Volume (Metric):
150 to 1500 μL
10 μL
20 μL
30 μL
50 μL
100 μL
200 μL
300 μL
1000 μL
5 mL
10 mL
Length (Metric):
3.2 cm
3.3 cm
4.8 cm
5.2 cm
6.99 cm
7.1 cm
7.8 cm
10.5 cm
14.7 cm
15 cm
16 product options available for selection
Product selection table with 16 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Length (Metric)
21377604 1000 μL 7.1 cm
21377354 10 μL 3.2 cm
2137801 20 μL 3.3 cm
21377387 50 μL 4.8 cm
21377169 10 μL 5.2 cm
21377353 20 μL 5.2 cm
1438775 30 μL 5.2 cm
21377170 100 μL 5.2 cm
21377171 200 μL 5.2 cm
1438654 100 μL 7.8 cm
21377804 200 μL 7.8 cm
21377307 300 μL 5.2 cm
21377605 1000 μL 10.5 cm
02681252 10 mL 15 cm
21377356 150 to 1500 μL 6.99 cm
21377512 5 mL 14.7 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 16 options available.
16 options
Catalog No. 21377604 Supplier Thermo Scientific™ Supplier No. 94052410
Edge

Protect pipettes and the samples from contamination with these filtered pipette tips.

Protect pipettes and the samples from contamination with Thermo Scientific™ Finntip™ Filtered Pipette Tips. These sterile, disposable pipette tips keep pipettes clean, and are perfect for PCR amplification applications and for pipetting radioactive, infectious, and aerosol-emitting samples. Available in racks with a wide volume range of 0.2μL-10mL.

Features:

  • Inert and nonsealing polyethylene filter
  • Tips come vacuum sealed in irradiation-sterilized hinged-tip racks
  • Not autoclavable
  • Certified to be free from DNA, DNase, RNase and endotoxin

Specifications

Description Finntip Filter 1000 μL, sterile
Volume (Metric) 1000 μL
Filtered Filtered
Sterility Sterile
Autoclavable Not Autoclavable
Length (Metric) 7.1 cm
Format Rack
Color Code Blue
Disposable Single-use
For Use With (Equipment) Thermo Scientific Finnpipette
Product Line FinnTip Filtered
Fit for Purpose Sterile & PCR Ready
Show More Show Less