Thermo Scientific™ Finnpipette™ F2 GLP Kit 4 PROMO
Eliminate the possible effects of hand warmth on the accuracy of measurements with these variable-volume pipettes, which feature a thermally-isolated AVG mechanism. Kit includes everything you need for use and enables you to cover the volume range you need.
Supplier:  Thermo Scientific™ P4701070
Catalog No. 05719621TI
Description
Description
  • AVG-volume gearing mechanism for accuracy and precision
  • Fully autoclavable for protection
  • Soft-touch tip ejector for light tip ejection
  • Large display for better vision ergonomics
  • Very light pipetting forces
Kit 4 includes:
  • Three Thermo Scientific™ Finnpipette Pipetters from 20 to 1000 μL:
    • Finnpipette F2
    • 2 to 20 μL, 20 to 200 μL
    • 100 to 1000 μL
  • Flex tips: (2) 200 μL, 10 x 96; 100 to 1000 μL, 10 x 96
  • F-stand
  • Good Laboratory Pipetting Guide
  • Reagent reservoir demo pack
Ordering Information:
The kit contains samples of Finntip Flex, the most advanced traditional tip offering very low attachment and ejection forces. The Good Laboratory Pipetting Guide contains all the information of pipetting from choosing the right type to detailed de-contamination information.
Warranty: 5 years with web registration.
Specifications
Specifications
200 and 1000 μL Flex Tips
Assorted
GLP Kit
Autoclavable
F2
2 to 1000 μL