missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cytiva Filter Holder

Polysulfone In-Line Filter Holder for 50mm Diameter Membrane

$153.68 - $1,849.00

Specifications

Diameter (Metric) Membrane 50 mm
View More Specs

Products 4
Catalog Number Mfr. No. Material Description Diameter (Metric) Membrane No. per Pack Price Quantity  
Catalog Number Mfr. No. Material Description Diameter (Metric) Membrane No. per Pack Price Quantity  
09927749
View Documents Promotion Details
Cytiva
10464100
Stainless Steel ML050/0 stainless steel 50 mm 1
Pack for $1,849.00
 
09927741
View Documents Promotion Details
Cytiva
10461300
Polysulfone FP050/1 PSU 50 mm 1
Pack for $155.61
 
09-927-740
View Documents Promotion Details
Cytiva
10461200
Polysulfone FP050/0 PSU 50 mm 5
Pack of 5 Each for $596.00
 
09927739
View Documents Promotion Details
Cytiva
10461100
Polysulfone FP050/0 PSU 50 mm 1
Pack for $153.68
 
Description

Description

Polysulfone In-Line Filter Holder for 50mm Diameter Membrane

Specifications

Specifications

50 mm
SDS
missing translation for 'documents'

missing translation for 'documents'