missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Synthware™ Filter Funnel
Funnel features a fritted disc and can be used to filter particulates from a variety of substances.
$152.15 - $198.82
Specifications
Material | Borosilicate Glass (3.3) |
---|---|
Color | Clear |
For Use With (Application) | Filtering Media |
Joint Number | 24/40 |
Porosity | Extra Coarse |
Description
- Filter funnel with a fritted disc
- Use to filter particulates from a variety of substances
Specifications
Borosilicate Glass (3.3) | |
Filtering Media | |
Extra Coarse |
Clear | |
24/40 | |
Filter Funnel |