Synthware™ Filter Funnel
Funnel features a fritted disc and can be used to filter particulates from a variety of substances.
Supplier: Synthware™ F932475X
Description
- Filter funnel with a fritted disc
- Use to filter particulates from a variety of substances
Specifications
75 mm | |
Clear | |
24/40 | |
Filter Funnel |
Borosilicate Glass (3.3) | |
Filtering Media | |
Extra Coarse |