missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Agilent BioTek Filter Cube, 377/647

Description
- Accessory for Agilent BioTek instruments range of microplate washers, readers and cell imagers
- Fluorescence filter
- Requires 365 nm LED cube (1225007)
Specifications
Specifications
| For Use With (Equipment) | Microplate readers and imagers |