missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Filter aid, Harborlite 800, Thermo Scientific Chemicals
$83.15 - $230.10
Specifications
Density | 2.3 |
---|---|
Chemical Name or Material | Filter aid, Harborlite 800 |
CAS | 130885-09-5 |
MDL Number | MFCD00197888 |
Synonym | Perlite, filter aid; Harborlite J2 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL1360836
|
Thermo Scientific Chemicals
L1360836 |
500 g |
Each for $83.15
|
|
|||||
AAL136080E
|
Thermo Scientific Chemicals
L136080E |
2500 g |
Each for $230.10
|
|
|||||
Description
Filter aid, Harborlite 800 is used in multiple industrial applications such as chemical processing, mineral and lubrication oil production, biodiesel purification. It is also used for filtration performance in brewing, juices, wine, sugar syrups, edible vegetable and fish oils.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsFilter aid, Harborlite 800 is used in multiple industrial applications such as chemical processing, mineral and lubrication oil production, biodiesel purification. It is also used for filtration performance in brewing, juices, wine, sugar syrups, edible vegetable and fish oils.
Solubility
Insoluble in water.
Notes
Harborlite is a registered trademark of Harborlite Corportion.
Specifications
2.3 | |
130885-09-5 | |
Perlite, filter aid; Harborlite J2 |
Filter aid, Harborlite 800 | |
MFCD00197888 | |
Insoluble in water. |
RUO – Research Use Only