missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Filter aid, Harborlite 800

Manufacturer:  Alfa Aesar™ L136080E

 View more versions of this product

Catalog No. AAL136080E

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Applications
Filter aid, Harborlite 800 is used in multiple industrial applications such as chemical processing, mineral and lubrication oil production, biodiesel purification. It is also used for filtration performance in brewing, juices, wine, sugar syrups, edible vegetable and fish oils.

Solubility
Insoluble in water.

Notes
Harborlite is a registered trademark of Harborlite Corportion.
Specifications

Specifications

Filter aid, Harborlite 800
2500g
MFCD00197888
Perlite, filter aid; Harborlite J2
Documents