missing translation for 'onlineSavingsMsg'
Learn More

Filter aid, Celite∣r 545, treated with sodium carbonate, flux calcined

Catalog No. p-7031898
Change view
Click to view available options
Quantity:
1000 g
5000 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH331520B 1000 g
AAH331520I 5000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH331520B Supplier Thermo Scientific Chemicals Supplier No. H331520B
Only null left

CAS: 68855-54-9 | CNa2O3 | 105.988 g/mol

Celite filter aids maximize biodiesel yields as a result of improved filtration, dewatering, dewaxing and winterization. A medium to fine product for the filtration of most process streams including beer, cider, wines, water, low viscosity glues and organic chemicals and pharmaceuticals. for water purification.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Celite filter aids maximize biodiesel yields as a result of improved filtration, dewatering, dewaxing and winterization. A medium to fine product for the filtration of most process streams including beer, cider, wines, water, low viscosity glues and organic chemicals and pharmaceuticals. for water purification.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong acids.

Chemical Identifiers

CAS 68855-54-9
Molecular Formula CNa2O3
Molecular Weight (g/mol) 105.988
MDL Number MFCD00132803
InChI Key CDBYLPFSWZWCQE-UHFFFAOYSA-L
PubChem CID 10340
ChEBI CHEBI:29377
IUPAC Name disodium;carbonate
SMILES C(=O)([O-])[O-].[Na+].[Na+]

Specifications

Quantity 1000 g
Solubility Information Insoluble in water.
Chemical Name or Material Filter aid, Celite™ 545, treated with sodium carbonate, flux calcined
Hazard Category Warning
Hazard Statement GHS H Statement
H373-H319-H335
May cause damage to organs through prolonged or repeated exposure.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P260-P261-P280-P305+P351+P338-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 272-489-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only