Learn More
Filter aid, Celite Hyflo Super-cel∣r, Thermo Scientific Chemicals
Filter aid, Celite Hyflo Super-cel, O2Si, CAS Number-68855-54-9, 500g, CHEBI:30563, 319-127-6, H319-H335-H350-H372
$76.52 - $208.18
Chemical Identifiers
CAS | 68855-54-9 |
---|---|
Molecular Formula | O2Si |
Molecular Weight (g/mol) | 60.083 |
MDL Number | MFCD00132803 |
InChI Key | VYPSYNLAJGMNEJ-UHFFFAOYSA-N |
PubChem CID | 24261 |
ChEBI | CHEBI:30563 |
IUPAC Name | dioxosilane |
SMILES | O=[Si]=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAB2265836
|
Thermo Scientific Chemicals
B2265836 |
500 g |
Each for $76.52
|
|
|||||
AAB226580E
|
Thermo Scientific Chemicals
B226580E |
2500 g |
Each for $208.18
|
|
|||||
Description
Filter aid, Celite Hyflo Super-cel™ is used as a filtration agent, a functional filler in paints, plastics, paper, rubber, adhesives, catalysts, agricultural chemicals, pharmaceuticals and other chemicals, and a mild abrasive in polishes and cleaners. It is also used in clarifying liquids and oils, as an efficient thermal insulator; in manufacture of heat insulators, fire brick, and fire- and acid-proof packing materials; as an absorbent; in absorbent dynamite; as a solid carrier in chromatography; pozzolanic admixture in concrete mixes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsFilter aid, Celite Hyflo Super-cel« is used as a filtration agent, a functional filler in paints, plastics, paper, rubber, adhesives, catalysts, agricultural chemicals, pharmaceuticals and other chemicals, and a mild abrasive in polishes and cleaners. It is also used in clarifying liquids and oils, as an efficient thermal insulator; in manufacture of heat insulators, fire brick, and fire- and acid-proof packing materials; as an absorbent; in absorbent dynamite; as a solid carrier in chromatography; pozzolanic admixture in concrete mixes.
Solubility
Insoluble in water.
Notes
Store in cool, dry conditions in a sealed container. Incompatible with acids.
Chemical Identifiers
68855-54-9 | |
60.083 | |
VYPSYNLAJGMNEJ-UHFFFAOYSA-N | |
CHEBI:30563 | |
O=[Si]=O |
O2Si | |
MFCD00132803 | |
24261 | |
dioxosilane |
Specifications
500 g | |
68855-54-9 | |
MFCD00132803 | |
Insoluble in water. | |
O=[Si]=O | |
60.083 | |
CHEBI:30563 |
Odorless | |
O2Si | |
14,4973 | |
VYPSYNLAJGMNEJ-UHFFFAOYSA-N | |
dioxosilane | |
24261 | |
Filter aid, Celite Hyflo Super-cel™ |
Safety and Handling
GHS H Statement
H350-H373-H319-H335
May cause cancer.
May cause damage to organs through prolonged or repeated exposure.
Causes serious eye irritation.
May cause respiratory irritation.
P201-P202-P260-P264b-P270-P271-P280i-P281-P304+P340-P305+P351+P338-P308+P313-P501c
H319-H335-H350-H372
EINECSNumber : 319-127-6
RTECSNumber : VV7311000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only