missing translation for 'onlineSavingsMsg'
Learn More

Filter aid, Celite Hyflo Super-cel∣r

Catalog No. AAB2265836
Change view
Click to view available options
Quantity:
500 g
2500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2265836 500 g
AAB226580E 2500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2265836 Supplier Thermo Scientific Chemicals Supplier No. B2265836

Filter aid, Celite Hyflo Super-cel, O2Si, CAS Number-68855-54-9, 500g, CHEBI:30563, 319-127-6, H319-H335-H350-H372 | CAS: 68855-54-9 | O2Si | 60.083 g/mol

Filter aid, Celite Hyflo Super-cel™ is used as a filtration agent, a functional filler in paints, plastics, paper, rubber, adhesives, catalysts, agricultural chemicals, pharmaceuticals and other chemicals, and a mild abrasive in polishes and cleaners. It is also used in clarifying liquids and oils, as an efficient thermal insulator; in manufacture of heat insulators, fire brick, and fire- and acid-proof packing materials; as an absorbent; in absorbent dynamite; as a solid carrier in chromatography; pozzolanic admixture in concrete mixes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Filter aid, Celite Hyflo Super-cel« is used as a filtration agent, a functional filler in paints, plastics, paper, rubber, adhesives, catalysts, agricultural chemicals, pharmaceuticals and other chemicals, and a mild abrasive in polishes and cleaners. It is also used in clarifying liquids and oils, as an efficient thermal insulator; in manufacture of heat insulators, fire brick, and fire- and acid-proof packing materials; as an absorbent; in absorbent dynamite; as a solid carrier in chromatography; pozzolanic admixture in concrete mixes.

Solubility
Insoluble in water.

Notes
Store in cool, dry conditions in a sealed container. Incompatible with acids.

Chemical Identifiers

CAS 68855-54-9
Molecular Formula O2Si
Molecular Weight (g/mol) 60.083
MDL Number MFCD00132803
InChI Key VYPSYNLAJGMNEJ-UHFFFAOYSA-N
PubChem CID 24261
ChEBI CHEBI:30563
IUPAC Name dioxosilane
SMILES O=[Si]=O

Specifications

Quantity 500 g
Odor Odorless
Merck Index 14,4973
Solubility Information Insoluble in water.
Chemical Name or Material Filter aid, Celite Hyflo Super-cel™
Hazard Category H319-H335-H350-H372
Hazard Statement GHS H Statement
H350-H373-H319-H335
May cause cancer.
May cause damage to organs through prolonged or repeated exposure.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P201-P202-P260-P264b-P270-P271-P280i-P281-P304+P340-P305+P351+P338-P308+P313-P501c
EINECSNumber 319-127-6
RTECSNumber VV7311000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only