missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Filter Aid, Celite AW Standard Supercel™

$71.72 - $71.72

Chemical Identifiers

CAS 61790-53-2
MDL Number MFCD00132803
Merck Index 14,4973
Synonym Celite 521AW, filter aid
Chemical Name or Material Filter Aid, Celite AW Standard Supercel™
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1455236 SDS Alfa Aesar™
L1455236
500g Each for $71.72
Specifications

Description

Applications
Filter Aid, Celite AW Standard Supercel« is used for general filtrations. It is also used as a functional filler in paints, plastics, paper, rubber, adhesives, catalysts, agricultural chemicals, pharmaceuticals and other chemicals, and a mild abrasive in polishes and cleaners. It is also used in clarifying liquids and oils, as an efficient thermal insulator; in manufacture of heat insulators, fire brick, and fire- and acid-proof packing materials; as an absorbent; in absorbent dynamite; as a solid carrier in chromatography; pozzolanic admixture in concrete mixes.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with acids.
Description & Specifications

Chemical Identifiers

61790-53-2
14,4973
Filter Aid, Celite AW Standard Supercel™
MFCD00132803
Celite 521AW, filter aid

Specifications

500g
MFCD00132803
61790-53-2
Filter Aid, Celite AW Standard Supercel™
Odorless
14,4973
Celite 521AW, filter aid

Safety and Handling

GHS H Statement
H350-H373-H319-H335
May cause cancer.
May cause damage to organs through prolonged or repeated exposure.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P260-P261-P280-P305+P351+P338-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

RTECSNumber : VV7311000

TSCA : TSCA