missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Filter aid, Celite™ 545, treated with sodium carbonate, flux calcined

Manufacturer:  Alfa Aesar™ H331520I

 View more versions of this product

Catalog No. AAH331520I



Description

Description

Applications
Celite filter aids maximize biodiesel yields as a result of improved filtration, dewatering, dewaxing and winterization. A medium to fine product for the filtration of most process streams including beer, cider, wines, water, low viscosity glues and organic chemicals and pharmaceuticals. for water purification.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong acids.
Specifications

Specifications

Filter aid, Celite™ 545
treated with sodium carbonate, flux calcined
CNa2O3
CDBYLPFSWZWCQE-UHFFFAOYSA-L
disodium;carbonate
10340
5000g
68855-54-9
MFCD00132803
C(=O)([O-])[O-].[Na+].[Na+]
105.988
CHEBI:29377

Chemical Identifiers

68855-54-9
105.988
CDBYLPFSWZWCQE-UHFFFAOYSA-L
CHEBI:29377
C(=O)([O-])[O-].[Na+].[Na+]
CNa2O3
MFCD00132803
10340
disodium;carbonate
Safety and Handling

Safety and Handling

GHS H Statement
H373-H319-H335
May cause damage to organs through prolonged or repeated exposure.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P260-P261-P280-P305+P351+P338-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA

SDS Documents