missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Honeywell Fluka™ Filter Agent, Celite™ 545, Honeywell™ Fluka™

Celite® 545

$121.50 - $121.50

Chemical Identifiers

CAS 68855-54-9
Molecular Formula CNa2O3
Molecular Weight (g/mol) 105.988
MDL Number MFCD00132803
InChI Key CDBYLPFSWZWCQE-UHFFFAOYSA-L
PubChem CID 10340
ChEBI CHEBI:29377
IUPAC Name disodium;carbonate
SMILES C(=O)([O-])[O-].[Na+].[Na+]
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Qty  
Catalog Number Mfr. No. Quantity Packaging Price Qty  
60046850 View Documents Honeywell Fluka™
419931500G
500g Poly Bottle Each for $121.50
 
Description

Description

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications

Chemical Identifiers

68855-54-9
105.988
CDBYLPFSWZWCQE-UHFFFAOYSA-L
CHEBI:29377
C(=O)([O-])[O-].[Na+].[Na+]
CNa2O3
MFCD00132803
10340
disodium;carbonate

Specifications

White to Yellow
68855-54-9
CNa2O3
NONH for all modes of transport
C(=O)([O-])[O-].[Na+].[Na+]
105.988
CHEBI:29377
500g
Poly Bottle
MFCD00132803
CDBYLPFSWZWCQE-UHFFFAOYSA-L
disodium;carbonate
10340
Filter agent
Safety and Handling

Safety and Handling

P260