Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Filamin Ab-1, Mouse Monoclonal Antibody


Ensure accurate, reproducible results in immunohistochemistry, western blotting and ELISA experiments with Thermo Scientific™ Filamin Ab-1, Mouse Monoclonal Antibody.

Manufacturer: thermo scientific™ lab vision™  MS1211P1ABX

 View more versions of this product

Catalog No. MS1211P1ABX

  • / Each
 

Description & Specifications

Specifications

Antigen Filamin Ab-1
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Classification Monoclonal
Clone FLMN01; same as PM6/317
Concentration 1mg/mL
Conjugate Unlabeled
Cross Reactivity Chicken
Cross Reactivity Guinea Pig
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rabbit
Cross Reactivity Rat
Format Concentrated
Formulation Ab without BSA/Azide (Frozen)
Host Species Murine
Immunogen Platelet filamin
Isotype IgG1
Quantity 100μL
Regulatory Status RUO
Research Discipline Cytoskeletal Protein
Type Primary
Monoclonal or Polyclonal Monoclonal
Target Species Chicken
Target Species Guinea Pig
Target Species Human
Target Species Murine
Target Species Rabbit
Target Species Rat

Ensure accurate, reproducible results in immunohistochemistry, western blotting and ELISA experiments with Thermo Scientific™ Filamin Ab-1, Mouse Monoclonal Antibody.

Filamin functions as a crosslinking protein forming a flexible link between two actin filaments. It is composed of two identical polypeptide chains each joined to the other at one end, with an actin binding site at the other. It is present in human platelets, lymphocytes, fibroblasts and smooth muscle actin.

Host Species: Mouse

Clone: FLMN01; same as PM6/317

Isotype: IgG1

Species Reactivity: Human, Mouse, Rabbit, Guinea Pig, Chicken and Rat. Others not known.

Epitope: Not determined

Immunogen: Platelet filamin

Molecular Weight: 250 kDa

Positive Control: NIH3T3 cells, Skin

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • ELISA

Note:

USA: RUO; Int'l: RUO