Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Filamin Ab-1, Mouse Monoclonal Antibody


Ensure accurate, reproducible results in immunohistochemistry, western blotting and ELISA experiments with Thermo Scientific™ Filamin Ab-1, Mouse Monoclonal Antibody.

Manufacturer: thermo scientific™ lab vision™  MS1211PABX

 View more versions of this product

Catalog No. MS1211PABX

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity Chicken
Cross Reactivity Guinea Pig
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rabbit
Cross Reactivity Rat
Antigen Filamin Ab-1
Immunogen Platelet filamin
Clone FLMN01; same as PM6/317
Regulatory Status RUO
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Format Concentrated
Isotype IgG1
Quantity 200µL
Host Species Mouse
Concentration 1mg/mL
Type Primary
Classification Monoclonal
Formulation Ab without BSA/Azide (Frozen)
Research Discipline Cytoskeletal Protein
Conjugate Unlabeled
Item Description 200uL; 1mg/mL; Purified w/o BSA/azide

Ensure accurate, reproducible results in immunohistochemistry, western blotting and ELISA experiments with Thermo Scientific™ Filamin Ab-1, Mouse Monoclonal Antibody.

Filamin functions as a crosslinking protein forming a flexible link between two actin filaments. It is composed of two identical polypeptide chains each joined to the other at one end, with an actin binding site at the other. It is present in human platelets, lymphocytes, fibroblasts and smooth muscle actin.

Host Species: Mouse

Clone: FLMN01; same as PM6/317

Isotype: IgG1

Species Reactivity: Human, Mouse, Rabbit, Guinea Pig, Chicken and Rat. Others not known.

Epitope: Not determined

Immunogen: Platelet filamin

Molecular Weight: 250 kDa

Positive Control: NIH3T3 cells, Skin

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • ELISA

Note:

USA: RUO; Int'l: RUO