Please login to your online account to display your discounted pricing

FIIN 1 hydrochloride, Tocris™

Potent, irreversible FGFR inhibitor

$290.95 - $1,205.80

Chemical Identifiers

Chemical Name or Material FIIN 1 hydrochloride
Assay Percent Range >96%
Molecular Formula C32H39Cl2N7O4
Formula Weight 693.06
Synonym N-(3-((3-(2,6-dichloro-3,5-dimethoxyphenyl)-7-(4-(diethylamino)butylamino)-2-oxo-3,4-dihydropyrimido[4,5-d]pyrimidin-1(2H)-yl)methyl)phenyl)acrylamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

400250

 
r & d systems
400250
50mg Each for $1,205.80

400210

 
r & d systems
400210
10mg Each for $290.95
Description & Specifications

Specifications

Formula Weight 693.06
Quantity 50mg
Chemical Name or Material FIIN 1 hydrochloride
Assay Percent Range >96%
Molecular Formula C32H39Cl2N7O4
CAS 1256152-35-8
Synonym N-(3-((3-(2,6-dichloro-3,5-dimethoxyphenyl)-7-(4-(diethylamino)butylamino)-2-oxo-3,4-dihydropyrimido[4,5-d]pyrimidin-1(2H)-yl)methyl)phenyl)acrylamide
Recommended Storage Store at -20°C
Safety and Handling

Store at -20°C