Thermo Scientific Chemicals Field's Stain A
Supplier: Thermo Scientific Chemicals A1857709
Description
Used to stain blood smears to identify malarial parasites
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Stain | |
10g |
RUO – Research Use Only