Please login to your online account to display your discounted pricing

Science First™ Field Map Kit

Group and compare lab plots and see representations of electrical fields.

$92.85 - $92.85

Specifications

For Use With (Application) Cartography training
View More Specs
Includes:  Cork board cover; 25 sheets black conductive paper, 20 × 25cm grid; 100 sheets white paper, 20 × 25cm grid; template; conductive ink pen; four connecting wires with alligator clip leads; illustrated instructions
Requires:  12VAC power supply or 1.5V battery, digital voltmeter (10 megohms or greater)
Products
Catalog Number Mfr. No. Product Type Price Quantity    

S63197

 
science first™
29-3000EMD
Conductive ink pen Each for $92.85
Description & Specifications

Specifications

For Use With (Application) Cartography training

  • Plot and compare electrical fields by drawing electrodes in conductive ink on semiconducting paper
  • Transcribe data to grid paper for a great group lab
  • Contains everything needed for a full year's use
  • Dimensions: 12.5 × 12 in. (31.8 × 30.5cm)