Please login to your online account to display your discounted pricing

Ficoll™ (Molecular Biology), MP Biomedicals


Quantity 25g

Manufacturer: mp biomedicals  0219482425

 View more versions of this product

Catalog No. ICN19482425

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 25g
Chemical Name or Material Ficoll™
Name Note Dialyzed
CAS 26873-85-8
Purity Grade Molecular Biology

Specifications

Chemical Name or Material Ficoll™
Name Note Dialyzed
CAS 26873-85-8
Purity Grade Molecular Biology