Please login to your online account to display your discounted pricing

Ficoll™ (Molecular Biology), MP Biomedicals

$249.75 - $249.75

Chemical Identifiers

Chemical Name or Material Ficoll™
Name Note Dialyzed
CAS 26873-85-8
Purity Grade Molecular Biology
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

ICN19482425

 
mp biomedicals
0219482425
25g Each for $249.75
Description & Specifications

Specifications

Quantity 25g
Chemical Name or Material Ficoll™
Name Note Dialyzed
CAS 26873-85-8
Purity Grade Molecular Biology