Please login to your online account to display your discounted pricing

Ficoll 400 (White to Off-White Powder/Molecular Biology), Fisher BioReagents


Quantity 10g
Packaging Poly Bottle

Manufacturer: fisher scientific company  FLBP52510

 View more versions of this product

Catalog No. BP525-10

  • / Each
  • / Case of 6 Each
 

Description & Specifications

Chemical Identifiers

Purity Grade Molecular Biology Grade
Physical Form Powder/Solid
Formula Weight ≈400,000
Quantity 10g
DNase DNase free
Protease Protease free
Viscosity [NULL]
Color Off-white
Packaging Poly Bottle
Specific Rotation Inclusive between 50° (+ or -)
Chemical Name or Material Ficoll™ 400
CAS 26873-85-8
ChemAlert Storage Symbol Gray
Recommended Storage RT

Specifications

Chemical Name or Material Ficoll™ 400
CAS 26873-85-8
Purity Grade Molecular Biology Grade
Physical Form Powder/Solid
Formula Weight ≈400,000
Accessories
 

RT

SDS & Safety Documents