missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Ficoll 400 (White to Off-White Powder/Molecular Biology), Fisher BioReagents™
$577.57 - $577.57

Chemical Identifiers
CAS | 26873-85-8 |
---|---|
Molecular Formula | C15H27ClO12 |
Molecular Weight (g/mol) | 434.819 |
InChI Key | IACFXVUNKCXYJM-AKSHDPDZSA-N |
Synonym | ficol 400 |
PubChem CID | 88330506 |
IUPAC Name | 2-(chloromethyl)oxirane;(2R,3R,4S,5S,6R)-2-[(2S,3S,4S,5R)-3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol |
SMILES | C1C(O1)CCl.C(C1C(C(C(C(O1)OC2(C(C(C(O2)CO)O)O)CO)O)O)O)O |
Description
Ficoll 400 (White to Off-White Powder/Molecular Biology)
Ficoll 400 is used as a component of Denhardt's Reagent and in gel-loading buffers for DNA gels. It is also used in forming density gradients to separate cells and subcellular components.Chemical Identifiers
26873-85-8 | |
434.819 | |
ficol 400 | |
2-(chloromethyl)oxirane;(2R,3R,4S,5S,6R)-2-[(2S,3S,4S,5R)-3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol |
C15H27ClO12 | |
IACFXVUNKCXYJM-AKSHDPDZSA-N | |
88330506 | |
C1C(O1)CCl.C(C1C(C(C(C(O1)OC2(C(C(C(O2)CO)O)O)CO)O)O)O)O |
Specifications
26873-85-8 | |
White | |
25g | |
IACFXVUNKCXYJM-AKSHDPDZSA-N | |
2-(chloromethyl)oxirane;(2R,3R,4S,5S,6R)-2-[(2S,3S,4S,5R)-3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol | |
88330506 | |
Powder/Solid | |
Poly Bottle | |
Inclusive between 50° (+ or -) |
C15H27ClO12 | |
ficol 400 | |
DNase free | |
C1C(O1)CCl.C(C1C(C(C(C(O1)OC2(C(C(C(O2)CO)O)O)CO)O)O)O)O | |
434.819 | |
≈400,000 | |
Molecular Biology | |
Protease free | |
Ficoll™ 400 |
Safety and Handling

Recommended Storage : RT