missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fibronectin Mouse anti-Human, Clone: FN-3, eBioscience™

Mouse Monoclonal Antibody

$117.48 - $262.68

Specifications

Antigen Fibronectin
Clone FN-3
Host Species Murine
Species Reactivity Human
Applications Immunohistochemistry (Formalin/Paraffin),Western Blot
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
50245608 Life Technologies
LS14986980
25μg Each for $117.48
50245609 Life Technologies
LS14986982
100μg Each for $262.68
Specifications

Description

The FN-3 monoclonal antibody recognizes human fibronectin, a component of the extracellular matrix. This high molecular weight protein (almost 400 kDa) exists as an extracellular and a soluble plasma form. Fibronectin functions in cell adhesion and migration via integrins present on the interacting cell. Fibronectin also associates with collagen, actin and fibrins. In malignancies, fibronectin protein levels can be altered. In lung carninomas, expression is decreased. The FN-3 antibody recognizes a determinant on human cellular but not plasma fibronectin; this recognition is not lost upon trypsin treatment. Furthermore, FN-3 antibody has been shown to crossreact to bovine fibronectin.
Description & Specifications

Specifications

Fibronectin
Murine
Immunohistochemistry (Formalin/Paraffin),Western Blot
Unlabeled
Store at 2-8°C.
Monoclonal
0.5mg/mL
FN-3
Human
RUO
Purified
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer