Please login to your online account to display your discounted pricing

Alfa Aesar™ Fibronectin, Human, 95%

For research use

$105.16 - $105.16

Specifications

Formulation Lyophilized powder
Recombinant No
Storage Requirements 2° to 8°C
Product Type Fibronectin, Human, 95%
Format Lyophilized powder
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ64560AMP

 
alfa aesar™
J64560AMP
1mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ64560MCR

 
alfa aesar™
J64560MCR
1mg Each for $105.16
Description & Specifications

Specifications

Formulation Lyophilized powder
Recombinant No
Storage Requirements 2° to 8°C
Product Type Fibronectin, Human, 95%
Format Lyophilized powder
For Use With (Application) Fibronectin is a major cell surface glycoprotein and is also present in blood plasma. Fibronectins are used to facilitate the attachment of cells to a variety of surfaces and to maintain cells in low serum conditions.
Purity 95%
Regulatory Status RUO