Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Fibronectin Ab-11, Mouse Monoclonal Antibody

Obtain accurate, reproducible results in immunofluorescence experiments with Thermo Scientific™ Fibronectin Ab-11, Mouse Monoclonal Antibody.

$274.18 - $1,168.80

Specifications

Antigen Fibronectin Ab-11
Clone FBN11
Applications Immunofluorescence
Classification Monoclonal
Conjugate Unlabeled
View More Specs
Products
Catalog Number Mfr. No. Concentration Quantity Price Quantity    

MS1351P

 
thermo scientific™ lab vision™
MS1351P
200μg/mL 1mL Each for $1,120.88

MS1351P0

 
thermo scientific™ lab vision™
MS1351P0
200μg/mL 100µL Each for $274.18

MS1351P1

 
thermo scientific™ lab vision™
MS1351P1
200μg/mL 500µL Each for $734.10

MS1351P1ABX

 
thermo scientific™ lab vision™
MS1351P1ABX
1mg/mL 100µL Each for $800.88

MS1351PABX

 
thermo scientific™ lab vision™
MS1351PABX
1mg/mL 200µL Each for $1,168.80
Description & Specifications

Specifications

Antigen Fibronectin Ab-11
Clone FBN11
Applications Immunofluorescence
Classification Monoclonal
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Format Concentrated
Host Species Mouse
Regulatory Status RUO
Research Discipline Extracellular Matrix
Type Primary
Immunogen Recombinant fibronectin protein
Isotype IgG1

Obtain accurate, reproducible results in immunofluorescence experiments with Thermo Scientific™ Fibronectin Ab-11, Mouse Monoclonal Antibody.

Fibronectin is a dimeric glycoprotein which is present in cells, extracellular matrix, and blood. It possesses at least four binding sites for collagen, glycosaminoglycans, transglutaminase, and a cell surface receptor. Fibronectin is involved in cell adhesion, tissue organization, and wound healing.

Host Species: Mouse

Clone: FBN11

Isotype: IgG1

Species Reactivity: Human, Mouse and Rat. Others not known.

Epitope: Not known

Immunogen: Recombinant fibronectin protein

Molecular Weight: 440 kDa (nonreduced) and 220 kDa (reduced)

Cellular Localization: Connective tissue matrix and basement membrane

Recommended for:

  • Immunofluorescence

Note:

USA: RUO; Int'l: RUO