Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Fibronectin Ab-11, Mouse Monoclonal Antibody


Obtain accurate, reproducible results in immunofluorescence experiments with Thermo Scientific™ Fibronectin Ab-11, Mouse Monoclonal Antibody.

Manufacturer: thermo scientific™ lab vision™  MS1351PABX

 View more versions of this product

Catalog No. MS1351PABX

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Antigen Fibronectin Ab-11
Format Concentrated
Host Species Mouse
Applications Immunofluorescence
Immunogen Recombinant fibronectin protein
Isotype IgG1
Quantity 200µL
Regulatory Status RUO
Clone FBN11
Research Discipline Extracellular Matrix
Type Primary
Classification Monoclonal
Conjugate Unlabeled
Concentration 1mg/mL

Obtain accurate, reproducible results in immunofluorescence experiments with Thermo Scientific™ Fibronectin Ab-11, Mouse Monoclonal Antibody.

Fibronectin is a dimeric glycoprotein which is present in cells, extracellular matrix, and blood. It possesses at least four binding sites for collagen, glycosaminoglycans, transglutaminase, and a cell surface receptor. Fibronectin is involved in cell adhesion, tissue organization, and wound healing.

Host Species: Mouse

Clone: FBN11

Isotype: IgG1

Species Reactivity: Human, Mouse and Rat. Others not known.

Epitope: Not known

Immunogen: Recombinant fibronectin protein

Molecular Weight: 440 kDa (nonreduced) and 220 kDa (reduced)

Cellular Localization: Connective tissue matrix and basement membrane

Recommended for:

  • Immunofluorescence

Note:

USA: RUO; Int'l: RUO