Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Fibronectin Ab-11, Mouse Monoclonal Antibody


Obtain accurate, reproducible results in immunofluorescence experiments with Thermo Scientific™ Fibronectin Ab-11, Mouse Monoclonal Antibody.

Manufacturer: thermo scientific™ lab vision™  MS1351P1ABX

 View more versions of this product

Catalog No. MS1351P1ABX

  • / Each
 

Description & Specifications

Specifications

Isotype IgG1
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Applications Immunofluorescence
Antigen Fibronectin Ab-11
Quantity 100µL
Immunogen Recombinant fibronectin protein
Format Concentrated
Regulatory Status RUO
Clone FBN11
Host Species Mouse
Concentration 1mg/mL
Type Primary
Classification Monoclonal
Conjugate Unlabeled
Research Discipline Extracellular Matrix

Obtain accurate, reproducible results in immunofluorescence experiments with Thermo Scientific™ Fibronectin Ab-11, Mouse Monoclonal Antibody.

Fibronectin is a dimeric glycoprotein which is present in cells, extracellular matrix, and blood. It possesses at least four binding sites for collagen, glycosaminoglycans, transglutaminase, and a cell surface receptor. Fibronectin is involved in cell adhesion, tissue organization, and wound healing.

Host Species: Mouse

Clone: FBN11

Isotype: IgG1

Species Reactivity: Human, Mouse and Rat. Others not known.

Epitope: Not known

Immunogen: Recombinant fibronectin protein

Molecular Weight: 440 kDa (nonreduced) and 220 kDa (reduced)

Cellular Localization: Connective tissue matrix and basement membrane

Recommended for:

  • Immunofluorescence

Note:

USA: RUO; Int'l: RUO