Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Fibronectin Ab-1, Mouse Monoclonal Antibody

Ensure accurate, reproducible results in western blotting, immunofluorescence, immunoprecipitation and biological blockade experiments.

$272.02 - $1,098.67

Specifications

Antigen Fibronectin Ab-1
Clone HFN 7.1
Applications Immunofluorescence
Applications Immunoprecipitation
Applications Western Blotting
View More Specs
Products
Catalog Number Mfr. No. Concentration Quantity Price Quantity    

MS165P

 
thermo scientific™ lab vision™
MS165P
200μg/mL 1mL Each for $1,002.88

MS165P0

 
thermo scientific™ lab vision™
MS165P0
200μg/mL 100µL Each for $272.02

MS165P1

 
thermo scientific™ lab vision™
MS165P1
200μg/mL 500µL Each for $665.18

MS165P1ABX

 
thermo scientific™ lab vision™
MS165P1ABX
1mg/mL 100µL Each for $762.59

MS165PABX

 
thermo scientific™ lab vision™
MS165PABX
1mg/mL 200µL Each for $1,098.67
Description & Specifications

Specifications

Antigen Fibronectin Ab-1
Clone HFN 7.1
Applications Immunofluorescence
Applications Immunoprecipitation
Applications Western Blotting
Applications Immunofluorescence
Applications Immunoprecipitation
Applications Western Blotting
Applications Immunofluorescence
Applications Immunoprecipitation
Applications Western Blotting
Classification Monoclonal
Conjugate Unlabeled
Cross Reactivity Human
Format Concentrated
Host Species Mouse
Regulatory Status RUO
Research Discipline Extracellular Matrix
Type Primary
Immunogen Human fibronectin purified from serum by affinity chromatography on gelatin-sepharose
Isotype IgG1

Ensure accurate, reproducible results in western blotting, immunofluorescence, immunoprecipitation and biological blockade experiments Thermo Scientific™ Fibronectin Ab-1, Mouse Monoclonal Antibody.

Fibronectin is a dimeric glycoprotein which is present in cells, extracellular matrix, and blood. It possesses at least four binding sites for collagen, glycosaminoglycans, transglutaminase, and a cell surface receptor. Fibronectin is involved in cell adhesion, tissue organization, and wound healing.

Host Species: Mouse

Clone: HFN 7.1

Isotype: IgG1

Species Reactivity: Human specific

Epitope: Peptide core

Immunogen: Human fibronectin purified from serum by affinity chromatography on gelatin-sepharose

Molecular Weight: 440 kDa (nonreduced) and 220 kDa (reduced)

Positive Control: SW156 cells

Cellular Localization: Connective tissue matrix and basement membrane

Recommended for:

  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation (Native and denatured)
  • Biological Blockade (Order Ab without azide)

Note:

USA: RUO; Int'l: RUO