Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Fibronectin Ab-1, Mouse Monoclonal Antibody


Ensure accurate, reproducible results in western blotting, immunofluorescence, immunoprecipitation and biological blockade experiments.

Manufacturer: thermo scientific™ lab vision™  MS165P1ABX

 View more versions of this product

Catalog No. MS165P1ABX

  • / Each
 

Description & Specifications

Specifications

Quantity 100µL
Host Species Mouse
Regulatory Status RUO
Cross Reactivity Human
Applications Immunofluorescence
Applications Immunoprecipitation
Applications Western Blotting
Immunogen Human fibronectin purified from serum by affinity chromatography on gelatin-sepharose
Format Concentrated
Antigen Fibronectin Ab-1
Isotype IgG1
Clone HFN 7.1
Concentration 1mg/mL
Classification Monoclonal
Conjugate Unlabeled
Research Discipline Extracellular Matrix
Type Primary

Ensure accurate, reproducible results in western blotting, immunofluorescence, immunoprecipitation and biological blockade experiments Thermo Scientific™ Fibronectin Ab-1, Mouse Monoclonal Antibody.

Fibronectin is a dimeric glycoprotein which is present in cells, extracellular matrix, and blood. It possesses at least four binding sites for collagen, glycosaminoglycans, transglutaminase, and a cell surface receptor. Fibronectin is involved in cell adhesion, tissue organization, and wound healing.

Host Species: Mouse

Clone: HFN 7.1

Isotype: IgG1

Species Reactivity: Human specific

Epitope: Peptide core

Immunogen: Human fibronectin purified from serum by affinity chromatography on gelatin-sepharose

Molecular Weight: 440 kDa (nonreduced) and 220 kDa (reduced)

Positive Control: SW156 cells

Cellular Localization: Connective tissue matrix and basement membrane

Recommended for:

  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation (Native and denatured)
  • Biological Blockade (Order Ab without azide)

Note:

USA: RUO; Int'l: RUO