missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fibroblast Growth Factor, basic, bovine, 95%, BT-105, Alfa Aesar™

Specifications

Format Lyophilized Powder
Packaging Plastic bottle
Purity 95%
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ65713YAM
View Documents
Thermo Fisher Scientific
J65713YAM
5μg N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAJ65713EXF
View Documents
Thermo Fisher Scientific
J65713EXF
25μg N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Specifications

Lyophilized Powder
Plastic bottle
95%
SDS