Please login to your online account to display your discounted pricing

Alfa Aesar™ Fibrinopeptide B, Human

For cellular and molecular biology applications

Specifications

Product Type Fibrinopeptide B, human
Cross Reactivity Human
Molecular Weight 1552.55
Storage Requirements -30° to -10°C
Format Solid
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ66812LB0

 
alfa aesar™
J66812LB0
5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ66812MCR

 
alfa aesar™
J66812MCR
1mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Product Type Fibrinopeptide B, human
Cross Reactivity Human
Molecular Weight 1552.55
Storage Requirements -30° to -10°C
Format Solid
Sequence Glp-Gly-Val-Asn-Asp-Asn-Glu-Glu-Gly-Phe-Phe-Ser-Ala-Arg
Regulatory Status RUO
Purification Method HPLC