Please login to your online account to display your discounted pricing

Alfa Aesar™ Fibrinopeptide B, Human


For cellular and molecular biology applications

Manufacturer: alfa aesar™  J66812MCR

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. AAJ66812MCR

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Specifications

Quantity 1mg
Format Solid
Sequence Glp-Gly-Val-Asn-Asp-Asn-Glu-Glu-Gly-Phe-Phe-Ser-Ala-Arg
Regulatory Status RUO
Storage Requirements -30° to -10°C
Product Type Fibrinopeptide B, human
Molecular Weight 1552.55
Purification Method HPLC
Cross Reactivity Human