Please login to your online account to display your discounted pricing

Alfa Aesar™ Fibrinogen Related Peptide

For cellular and molecular biology applications

Specifications

Product Type Fibrinogen Related Peptide
Molecular Weight 1502.59
Storage Requirements -30° to -10°C
Sequence Gly-Gln-Gln-His-His-Leu-Gly-Gly-Ala-Lys-Gln-Ala-Gly-Asp-Val
Format Solid
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ66069LB0

 
alfa aesar™
J66069LB0
5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ66069MA

 
alfa aesar™
J66069MA
10mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Product Type Fibrinogen Related Peptide
Molecular Weight 1502.59
Storage Requirements -30° to -10°C
Sequence Gly-Gln-Gln-His-His-Leu-Gly-Gly-Ala-Lys-Gln-Ala-Gly-Asp-Val
Format Solid
Regulatory Status RUO
Purification Method HPLC