Please login to your online account to display your discounted pricing

Alfa Aesar™ Fibrinogen-Binding Inhibitor Peptide

For cellular and molecular biology applications

Specifications

Product Type Fibrinogen-Binding Inhibitor Peptide
Molecular Weight 1189.30
Sequence H-His-His-Leu-Gly-Gly-Ala-Lys-Gln-Ala-Gly-Asp-Val-OH
For Use With (Application) Inhibits the binding of fibrinogen and fibronectin
Format Solid
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ60863LB0

 
alfa aesar™
J60863LB0
5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ60863MCR

 
alfa aesar™
J60863MCR
1mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Product Type Fibrinogen-Binding Inhibitor Peptide
Molecular Weight 1189.30
Sequence H-His-His-Leu-Gly-Gly-Ala-Lys-Gln-Ala-Gly-Asp-Val-OH
For Use With (Application) Inhibits the binding of fibrinogen and fibronectin
Format Solid
Common Name Fibrinogen γ-chain dodecapeptide
Regulatory Status RUO
Purification Method HPLC