Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Fibrillin-1 Ab-1, Mouse Monoclonal Antibody

Ensure accurate, reproducible results in immunohistochemistry, western blotting, immunofluorescence and electron microscopy experiments.

$272.53 - $1,227.29

Specifications

Antigen Fibrillin-1 Ab-1
Clone 11C1.3
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Western Blotting
View More Specs
Products
Catalog Number Mfr. No. Concentration Quantity Price Quantity    

MS231P

 
thermo scientific™ lab vision™
MS231P
200μg/mL 1mL Each for $1,147.61

MS231P0

 
thermo scientific™ lab vision™
MS231P0
200μg/mL 100μL Each for $272.53

MS231P1

 
thermo scientific™ lab vision™
MS231P1
200μg/mL 500μL Each for $763.78

MS231P1ABX

 
thermo scientific™ lab vision™
MS231P1ABX
1mg/mL 100μL Each for $848.41

MS231PABX

 
thermo scientific™ lab vision™
MS231PABX
1mg/mL 200μL Each for $1,227.29
Description & Specifications

Specifications

Antigen Fibrillin-1 Ab-1
Clone 11C1.3
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Western Blotting
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Western Blotting
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Western Blotting
Classification Monoclonal
Conjugate Unlabeled
Cross Reactivity Bovine
Cross Reactivity Human
Cross Reactivity Bovine
Cross Reactivity Human
Format Concentrated
Host Species Murine
Regulatory Status RUO
Research Discipline Extracellular Matrix
Type Primary
Immunogen Microfibrils from zonular apparatus of bovine eye
Isotype IgG2a kappa

Ensure accurate, reproducible results in immunohistochemistry, western blotting, immunofluorescence and electron microscopy experiments with Thermo Scientific™ Fibrillin-1 Ab-1, Mouse Monoclonal Antibody.

Fibrillin-1 is an extracellular acidic protein with a high cysteine content and an extended thread-like shape with mosaic composition of different types of extracellular modules. Most of the fibrillin molecule is contributed by 47 epidermal growth factor-like (EGF-like) repeats; 43 of them have a consensus sequence for calcium binding (cb). Other modules including 8-cysteine motifs (seven), hybrid motifs (two), and unique domains (three) are interspersed throughout the molecule[3-7]. fibrillin-1 contains a proline-rich region close to the N-terminus that is replaced by a glycine-rich region in fibrillin-2. The fibrillins are found throughout the connective tissue as integral components of extended fibrils.

Host Species: Mouse

Clone: 11C1.3

Isotype: IgG1/κ

Species Reactivity: Human and Cow. Others not known.

Epitope: aa 451-909

Immunogen: Microfibrils from zonular apparatus of bovine eye

Molecular Weight: 350 kDa

Positive Control: Normal skin fibroblasts

Cellular Localization: Extracellular microfibrils

Recommended for:

  • Immunohistochemistry (Acetone-fixed frozen)
  • Western Blotting
  • Immunofluorescence
  • Electron Microscopy

Note:

USA: RUO; Int'l: RUO