Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Fibrillin-1 Ab-1, Mouse Monoclonal Antibody


Ensure accurate, reproducible results in immunohistochemistry, western blotting, immunofluorescence and electron microscopy experiments.

Manufacturer: thermo scientific™ lab vision™  MS231P1ABX

 View more versions of this product

Catalog No. MS231P1ABX

  • / Each
 

Description & Specifications

Specifications

Regulatory Status RUO
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Western Blotting
Isotype IgG2a kappa
Host Species Mouse
Immunogen Microfibrils from zonular apparatus of bovine eye
Cross Reactivity Bovine
Cross Reactivity Human
Antigen Fibrillin-1 Ab-1
Format Concentrated
Quantity 100µL
Clone 11C1.3
Research Discipline Extracellular Matrix
Conjugate Unlabeled
Type Primary
Classification Monoclonal
Concentration 1mg/mL

Ensure accurate, reproducible results in immunohistochemistry, western blotting, immunofluorescence and electron microscopy experiments with Thermo Scientific™ Fibrillin-1 Ab-1, Mouse Monoclonal Antibody.

Fibrillin-1 is an extracellular acidic protein with a high cysteine content and an extended thread-like shape with mosaic composition of different types of extracellular modules. Most of the fibrillin molecule is contributed by 47 epidermal growth factor-like (EGF-like) repeats; 43 of them have a consensus sequence for calcium binding (cb). Other modules including 8-cysteine motifs (seven), hybrid motifs (two), and unique domains (three) are interspersed throughout the molecule[3-7]. fibrillin-1 contains a proline-rich region close to the N-terminus that is replaced by a glycine-rich region in fibrillin-2. The fibrillins are found throughout the connective tissue as integral components of extended fibrils.

Host Species: Mouse

Clone: 11C1.3

Isotype: IgG1/κ

Species Reactivity: Human and Cow. Others not known.

Epitope: aa 451-909

Immunogen: Microfibrils from zonular apparatus of bovine eye

Molecular Weight: 350 kDa

Positive Control: Normal skin fibroblasts

Cellular Localization: Extracellular microfibrils

Recommended for:

  • Immunohistochemistry (Acetone-fixed frozen)
  • Western Blotting
  • Immunofluorescence
  • Electron Microscopy

Note:

USA: RUO; Int'l: RUO