Fiberglass Fire Blanket in Packet
Lightweight and stored in a highly visible packet.
Manufacturer: Fisher Science Education BRID6X4
Description
- Smother fires, wrap a burn victim or shield against encroaching flames
- Stored in a highly visible, red vinyl packet that opens easily
- Woven glass fiber material is totally safe to handlers
- Stitched with flame-proof thread
Specifications
6 ft. x 4 ft. (1.8m x 1.2m) | |
1.8m | |
1.2m | |
Smother fires, wrap a burn victim or shield against encroaching flames |
6 ft. | |
4 ft. | |
Red | |
Fiberglass |