missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fiberglass Fire Blanket in Packet

Lightweight and stored in a highly visible packet.

$126.33 - $176.83

Specifications

Color Red
For Use With (Application) Smother fires, wrap a burn victim or shield against encroaching flames
Material Fiberglass
View More Specs

Products 2
Catalog Number Mfr. No. Length (English) Length (Metric) Width (English) Width (Metric) Price Quantity  
Catalog Number Mfr. No. Length (English) Length (Metric) Width (English) Width (Metric) Price Quantity  
S47560B
View Documents
Fisher Science Education
BRID6X4
6 ft. 1.8m 4 ft. 1.2m
Each for $126.33
 
S47560C
View Documents
Fisher Science Education
BRID6X6
6 ft. 1.8m 6 ft. 1.8m
Each for $176.83
 
Description

Description

  • Smother fires, wrap a burn victim or shield against encroaching flames
  • Stored in a highly visible, red vinyl packet that opens easily
  • Woven glass fiber material is totally safe to handlers
  • Stitched with flame-proof thread

Specifications

Specifications

Red
Smother fires, wrap a burn victim or shield against encroaching flames
Fiberglass
SDS